missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzenesulfonic Acid Monohydrate 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 500 g |
| UN Number | 2585 |
| Formula Weight | 158.17 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Benzenesulfonic Acid Monohydrate |
Safety and Handling
| EINECSNumber | (3)-1928 |
| RTECSNumber | DB4200000 |
| TSCA | Yes |