missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonamide 98.0+%, TCI America™

Catalog No. B0028500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0028500G 500 g
B002825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0028500G Supplier TCI America Supplier No. B0028500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-10-2
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.187
MDL Number MFCD00007930
InChI Key KHBQMWCZKVMBLN-UHFFFAOYSA-N
Synonym benzenesulphonamide, benzosulfonamide, benzolsulfonamide, phenyl sulfonamide, benzene sulfonamide, phenylsulfonamide, m and b 7973, chembl27601, benzolsulfonamid, benzensulfonamide
PubChem CID 7370
IUPAC Name benzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)N

Specifications

Melting Point 154°C
Color White
Quantity 500 g
Formula Weight 157.19
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Benzenesulfonamide
EINECSNumber (3)-3130
RTECSNumber DA9380000
TSCA Yes