missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfohydroxamic Acid 98.0+%, TCI America™

Catalog No. H02035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02035G 5 g
H020325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02035G Supplier TCI America Supplier No. H02035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 599-71-3
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
MDL Number MFCD00025013
InChI Key BRMDATNYMUMZLN-UHFFFAOYSA-N
Synonym piloty's acid, benzenesulfohydroxamic acid, benzenesulfonamide, n-hydroxy, benzenesulphonohydroxamic acid, benzsulfohydroxamic acid, hydroxamic acid, benzsulfo, n-phenylsulfonyl hydroxylamine, hydroxylamine, n-phenylsulfonyl, unii-c64l4umh19, n-hydroxy-benzenesulfonamide
PubChem CID 69033
IUPAC Name N-hydroxybenzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)NO

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 5 g
Formula Weight 173.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzenesulfohydroxamic Acid
RTECSNumber MX9350000
TSCA Yes