missing translation for 'onlineSavingsMsg'
Learn More

Benzeneboronic acid, 98+%

Catalog No. AAA1425709
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1425709 10 g
AAA1425718 50 g
AAA1425730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1425709 Supplier Thermo Scientific Chemicals Supplier No. A1425709

CAS: 98-80-6 | C6H7BO2 | 121.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Benzeneboronic acid is widely utilized in various cross coupling reactions for example Mizoroki-Heck and Suzuki-Miyaura coupling reactions, etc. It serves as a receptor for carbohydrates, antimicrobial agents and enzyme inhibitors in biological reactions. It acts as a reagent in palladium-catalyzed stereoselective Heck-type reaction and as a catalyst in Ni(II) pincer complex and Pd(II) pyridoxal hydrazone metallacycles. It is also employed as a sensor for neutron capture therapy for cancer, transmembrane transport and bioconjugation and labeling of proteins and cell surface.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Avoid moisture exposure.

Chemical Identifiers

CAS 98-80-6
Molecular Formula C6H7BO2
Molecular Weight (g/mol) 121.93
MDL Number MFCD00002103
InChI Key HXITXNWTGFUOAU-UHFFFAOYSA-N
Synonym benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2
PubChem CID 66827
ChEBI CHEBI:44923
IUPAC Name phenylboronic acid
SMILES OB(O)C1=CC=CC=C1

Specifications

Melting Point 214°C to 219°C
Quantity 10 g
Beilstein 970972
Sensitivity Hygroscopic
Merck Index 14,1068
Solubility Information Slightly soluble in water.
Formula Weight 121.93
Percent Purity ≥98%
Chemical Name or Material Benzeneboronic acid
Hazard Category H302
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264-P270-P301+P312-P304+P312-P305+P351+P338-P312-P330-P501c
EINECSNumber 202-701-9
RTECSNumber CY8575000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only