missing translation for 'onlineSavingsMsg'
Learn More

Benzene-1,3-disulfonic acid disodium salt, 94%

Catalog No. AAB2281136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2281136 500 g
AAB2281114 25 g
AAB2281122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2281136 Supplier Thermo Scientific Chemicals Supplier No. B2281136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 831-59-4 | C6H4Na2O6S2 | 282.19 g/mol

Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.

Solubility
Soluble in water( 663 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 831-59-4
Molecular Formula C6H4Na2O6S2
Molecular Weight (g/mol) 282.19
MDL Number MFCD00007492
InChI Key XWPWZOJBTOJEGW-UHFFFAOYSA-L
Synonym disodium 1,3-benzenedisulfonate, disodium benzene-1,3-disulfonate, 1,3-benzenedisulfonic acid, disodium salt, benzene-1,3-disulfonic acid disodium salt, 1,3-benzenedisulfonic acid disodium salt, disodium benzene-1,3-disulphonate, disodium m-benzenedisulfonate, sodium benzene-1,3-disulfonate, 1,3-benzenedisulfonic acid, sodium salt 1:2, m-benzenedisulfonic acid, disodium salt
PubChem CID 71577
IUPAC Name disodium;benzene-1,3-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1)S([O-])(=O)=O

Specifications

Density 1.124
Quantity 500 g
Solubility Information Soluble in water( 663g/L at 20°C).
Formula Weight 282.2
Percent Purity 94%
Chemical Name or Material Benzene-1,3-disulfonic acid disodium salt
EINECSNumber 212-606-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only