missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzanthrone 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 176°C |
| Quantity | 25 g |
| Formula Weight | 230.27 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzanthrone |
Sécurité et manipulation
| EINECSNumber | (4)-0793 |
| RTECSNumber | CX5075000 |
| TSCA | Yes |