missing translation for 'onlineSavingsMsg'
Learn More

Benzamide 99.0+%, TCI America™

Catalog No. B001225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B001225G 25 g
B0012500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B001225G Supplier TCI America Supplier No. B001225G

Chemical Identifiers

CAS 55-21-0
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00007968
InChI Key KXDAEFPNCMNJSK-UHFFFAOYSA-N
Synonym benzoic acid amide, benzoylamide, phenylcarboxyamide, phenylcarboxamide, benzenecarboxamide, amid kyseliny benzoove, amid kyseliny benzoove czech, phenyl carboxyamide, ccris 4594, benzoic acid,amide
PubChem CID 2331
ChEBI CHEBI:28179
IUPAC Name benzamide
SMILES C1=CC=C(C=C1)C(=O)N

Specifications

Melting Point 128°C
Color White
Boiling Point 288°C
Quantity 25 g
Formula Weight 121.14
Percent Purity ≥99.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Benzamide
RTECSNumber CU8700000
TSCA Yes