missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Boiling Point | >100.0°C |
| CAS Min % | 49.0, 48.0 |
| CAS Max % | 52.0, 51.0 |
| Linear Formula | C6H5CH2N(CH3)2RClR=C8H17toC18H37 |
| Concentration | 49 to 52% (based on M.W.= 352.5) |
| Density | 0.9800g/mL |
| Merck Index | 15, 1061 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone, slightly soluble in benzene, almost insoluble in ether |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS P Statement Causes severe skin burns and eye damage. Harmful if swallowed. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid release to the environment. |
RUO – Research Use Only