missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride (50% in Water), TCI America™

Catalog No. B0414500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0414500G 500 g
B041425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0414500G Supplier TCI America Supplier No. B0414500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63449-41-2
Molecular Formula C21H38ClN
Molecular Weight (g/mol) 339.99
MDL Number MFCD00137276
InChI Key JBIROUFYLSSYDX-UHFFFAOYSA-M
Synonym benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides
PubChem CID 13762
IUPAC Name benzyl(dodecyl)dimethylazanium chloride
SMILES [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Color Yellow
Quantity 500 g
UN Number 1760
Physical Form Liquid
Chemical Name or Material Benzalkonium Chloride (50% in Water)
EINECSNumber (3)-2694
RTECSNumber BO3151000
TSCA Yes