missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride 50% Solution, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019397
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019397 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019397 Supplier Merck Emd Millipore Supplier No. PHR16811ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Benzalkonium chloride is an antimicrobial preservative and is widely used against Gram-positive and Gram-negative organisms. It finds applications in cosmetic industry and pharmaceutical formulations.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
CAS 63449-41-2
MDL Number MFCD00145757
Synonym Benzalkonium chloride solution; Alkylbenzyldimethylammonium chloride
Quantity 1 mL
Grade Certified Reference Material