missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride, 17% (w/v), Solution, Spectrum™ Chemical

Catalog No. 18600879
Change view
Click to view available options
Quantity:
500 mL
4 L
200 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600879 500 mL Amber Glass Bottle
18600880 4 L Amber Glass Bottle
18600881 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600879 Supplier Spectrum Chemical Mfg Cor Supplier No. B1069500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

B1069, 8001-54-5

Spectrum™ Chemical Benzalkonium Chloride, 17 Percent (w/v), Solution is also referred to as BZK is an aqueous solution for handling purposes. It varies in color, from colorless to a pale yellow and has an almond-like smell and bitter taste. Its uses vary from disinfectant to microbial corrosion inhibition and can also be used as an algaecide.

Chemical Identifiers

CAS 8001-54-5 , 7732-18-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
SMILES [Cl-].C[N+](C)(*)CC1=CC=CC=C1

Specifications

CAS Max % 17%, 83%
Packaging Amber Glass Bottle
Quantity 500 mL
UN Number UN1760
Percent Purity 17% ±1.5
Grade Reagent
Physical Form Liquid