missing translation for 'onlineSavingsMsg'
Learn More

Kinematica Inc Benchtop Autoclave Sterilizer

Catalog No. 12983544
Change view
Click to view available options
Capacity (Metric):
3 L
30 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12983544 3 L
12983543 30 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12983544 Supplier Kinematica Inc Supplier No. PRIME3S

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diversified Applications : Agar Preparation, Liquid Sterilization, Empty Glassware Sterilization, Instrument and Solid Sterilization, Waste Liquid / Solid / Mixed Sterilization, Respirator Mask Sterilization,Other Heat Resistant Materials.

  • Features:
  • Advanced Steri-Touch Panel HMI
  • High-Accuracy Temperature Control
  • Integrated Steam generator
  • Flexible Load and Personnel Management
  • Powerful Vacuum System
  • Comprehensive Filtration System
  • Fully Customizable Programs
  • F0 Value Calculation
  • Includes Helix
  • Bowie-Dick
  • Ample 2G Data Storage
  • Automatic Water Management
  • Integrated Digital Recorder
  • Thermal Printer

Specifications

Autoclavable Autoclavable
Capacity (English) 0.79 gal.
Capacity (Metric) 3 L
Depth (Metric) Chamber 380 mm
Depth (English) Chamber 15 in.
Length (English) 9.3 in.
Length (Metric) 237 mm
Product Type Benchtop Autoclave
Temperature (English) 249°F to 275°F
Temperature (Metric) 121°C to 134°C
Latex Free Yes
Sterility Sterile
Show More Show Less