missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Bulb Pipettor

Catalog No. 1420954A
Change view
Click to view available options
Number of Channels:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels
1420954A 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1420954A Supplier Bel-Art™ Supplier No. H37888 0000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bulb pipettor is an ideal aspirator for use with Westergren or Wintrobe sedimentation tubes. Allows users to keep the bulb with a reagant bottle.

  • Comes complete with an elastic cord for dedicating that pipettor to a specific reagent bottle
  • Tapered, silicone rubber seal provides an airtight fit on all size pipettes
  • Can be opened, cleaned and steam autoclaved at 121ºC (250ºF)

Specifications

Autoclavable Autoclavable
Color Red
Disposable Reusable
For Use With Pipette
Grip Type Round
Material Rubber
Model Standard
Number of Channels 1
Sterility Non-sterile
Type Bulb Pipettor
For Use With (Equipment) Westergren and Wintrobe sedimentation tubes
Includes Elastic Cord
Show More Show Less