missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Sterileware™ Sampling Spatula, 14 in.
Description
Time-honored V-shape makes transferring powdered, flaked, crystalline or granular media a breeze
Long, 14 in. spatula is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 0.75 in. (18mm)
- 14 in. length
- High-impact polystyrene
- Individually packaged
- Sterilized by gamma irradiation
- Lot stamped, shipped with Certificate of Processing
- Sterile 9-in. model is available, 14-358-20
- Nonsterile 9-in. model: 03-421-149
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Shape | Pointed End |
| Sterility | Sterile |
| Capacity (Metric) | 23 mL |
| Disposable | Single-use |
| Product Type | Sampling Spatula with V-shaped channel |
| For Use With (Equipment) | Tall bottles or vessels |
| For Use With (Application) | Ideal for every type of powdered or granulated material, perfect for taking sterile samples during process quality inspections, dislodge caked or crystallized reagents from deep within tall bottles or vessels |
| Length (English) Blade | 10 in. |
| Length (English) | 14 in. |
| Show More |