missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Stainless-Steel Reagent Digger

Catalog No. 11865141
Change view
Click to view available options
Length (Metric):
22.9 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11865141 22.9 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11865141 Supplier Bel-Art™ Supplier No. F367130000
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ergonomically designed, time-tested tool, based on confirmed geometry

  • Dislodge stubborn, caked, or crystallized reagents from deep down inside a bottle or vessel that has a minimum neck diameter of 24 mm.
  • Shape facilitates scooping, shoveling, digging and measuring of all powdered, flaked, crystalline or granular media.

Specifications

Product Type Reagent Digger
Material Stainless Steel
Autoclavable Not Autoclavable
For Use With (Application) Scooping, shoveling, digging and measuring of all powdered, flaked, crystalline or granular media from deep down inside a bottle or vessel that has a minimum neck diameter of 24mm
For Use With (Equipment) 2.4cm dia. bottles
Length (English) 9 in.
Length (Metric) 22.9 cm