Learn More
Bel-Art™ Right-to-Know Safety-Vented Wash Bottles, GHS Labeling
Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips. Allows dispensing of wash bottle in upright or inverted position.
$133.29 - $223.46
Specifications
| Material | LDPE |
|---|---|
| Sterility | Non-sterile |
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Catalog Number | Mfr. No. | Capacity (Metric) | Chemical Label | Color | Quantity | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Chemical Label | Color | Quantity | Price | Quantity & Availability | ||
|
03411720
|
Bel-Art™
F12416 0001 |
500 mL | Acetone | Red | 4/Pk. |
Pack of 4 Each for $180.00
Case of 6 Pack for $1,080.00
|
|
||
|
03411726
|
Bel-Art™
F124160010 |
500 mL | Machine Oil | Natural | 4/Pk. |
Pack of 4 Each for $213.54
Case of 6 Pack for $1,281.24
|
|
||
|
03411722
|
Bel-Art™
F124160003 |
500 mL | Deionized Water | Blue | 4/Pk. |
Pack of 4 Each for $179.68
Case of 6 Pack for $1,078.08
|
|
||
|
03411724
|
Bel-Art™
F124160007 |
500 mL | Ethyl Acetate | Green | 4/Pk. |
Pack of 4 Each for $209.82
Case of 6 Pack for $1,258.92
|
|
||
|
03411731
|
Bel-Art™
F124160016 |
500 mL | Toluene | Red | 4/Pk. |
Pack of 4 Each for $223.46
Case of 6 Pack for $1,340.76
|
|
||
|
03411721
|
Bel-Art™
F124160002 |
500 mL | Dichloromethane | Yellow | 4/Pk. |
Pack of 4 Each for $180.04
Case of 6 Pack for $1,080.24
|
|
||
|
03411723
|
Bel-Art™
F124160004 |
500 mL | Distilled Water | Blue | 4/Pk. |
Pack of 4 Each for $180.04
Case of 6 Pack for $1,080.24
|
|
||
|
03411725
|
Bel-Art™
F124160008 |
500 mL | Isopropanol | Yellow | 4/Pk. |
Pack of 4 Each for $180.04
Case of 6 Pack for $1,080.24
|
|
||
|
03411729
|
Bel-Art™
F124160013 |
500 mL | Isotonic Saline | Natural | 4/Pk. |
Pack of 4 Each for $209.82
Case of 6 Pack for $1,258.92
|
|
||
|
03411727
|
Bel-Art™
F124160011 |
500 mL | Methanol | Green | 4/Pk. |
Pack of 4 Each for $180.04
Case of 6 Pack for $1,080.24
|
|
||
|
03411730
|
Bel-Art™
F124160015 |
500 mL | Sodium Hypochlorite (Bleach) | Yellow | 4/Pk. |
Pack of 4 Each for $180.04
Case of 6 Pack for $1,080.24
|
|
||
|
03411732
|
Bel-Art™
F124160017 |
500 mL | Water | Blue | 4/Pk. |
Pack of 4 Each for $191.75
Case of 6 Pack for $1,150.50
|
|
||
|
03411733
|
Bel-Art™
F124160019 |
500 mL | Ethanol | Natural | 4/Pk. |
Pack of 4 Each for $179.93
Case of 6 Pack for $1,079.58
|
|
||
|
03411734
|
Bel-Art™
F124160050 |
500 mL | Assorted | Assorted | 4/Pk. |
Pack of 4 Each for $223.46
Case of 6 Pack for $1,340.76
|
|
||
|
03411728
|
Bel-Art™
F124160012 |
500 mL | Methyl Ethyl Ketone | Green | 4/Pk. |
Pack of 4 Each for $180.04
Case of 6 Pack for $1,080.24
|
|
||
|
03411738
|
Bel-Art™
F124320011 |
1000 mL | Methanol | Green | 2/Pk. |
Pack of 2 Each for $165.46
Case of 6 Pack for $992.76
|
|
||
|
03411735
|
Bel-Art™
F124320001 |
1000 mL | Acetone | Red | 2/Pk. |
Pack of 2 Each for $133.29
Case of 6 Pack for $799.74
|
|
||
|
03411737
|
Bel-Art™
F124320008 |
1000 mL | Isopropanol | Yellow | 2/Pk. |
Pack of 2 Each for $165.46
Case of 6 Pack for $992.76
|
|
||
|
03411736
|
Bel-Art™
F124320004 |
1000 mL | Distilled Water | Blue | 2/Pk. |
Pack of 2 Each for $133.29
Case of 6 Pack for $799.74
|
|
||
|
03411740
|
Bel-Art™
F124320019 |
1000 mL | Ethanol | Natural | 2/Pk. |
Pack of 2 Each for $165.46
Case of 6 Pack for $992.76
|
|
||
|
03411739
|
Bel-Art™
F124320015 |
1000 mL | Sodium Hypochlorite (Bleach) | Yellow | 2/Pk. |
Pack of 2 Each for $165.46
Case of 6 Pack for $992.76
|
|
||
Description
Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position
- Wide mouth 53 mm (2 in.) for easy refilling
- Translucent LDPE bottles with color-coded caps for easy identification
- Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word
- Methyl Ethyl Ketone and Toluene do not include vented valve
- Assortment is 4 per bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol
- Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
- Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word.
Compliance
Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to Workplace Labels
Specifications
| LDPE | |
| Yes | |
| No | |
| Not Autoclavable |
| Non-sterile | |
| Reusable | |
| Cylindrical |