missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Right-to-Know Safety-Vented Wash Bottles, GHS Labeling
Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips. Allows dispensing of wash bottle in upright or inverted position.
Supplier: Bel-Art™ F124320004
Description
Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position
- Wide mouth 53 mm (2 in.) for easy refilling
- Translucent LDPE bottles with color-coded caps for easy identification
- Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word
- Methyl Ethyl Ketone and Toluene do not include vented valve
- Assortment is 4 per bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol
- Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
- Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word.
Compliance
Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to Workplace Labels
Chemical Identifiers
| LDPE | |
| Yes | |
| No | |
| Not Autoclavable |
| Non-sterile | |
| Reusable | |
| Cylindrical |
Specifications
| 32 oz., 1000 mL | |
| 32 oz. | |
| LDPE | |
| Non-sterile | |
| Reusable | |
| Wide, Threaded | |
| Safety-vented Wash Bottle | |
| Not Autoclavable |
| 1000 mL | |
| Blue | |
| 2/Pk. | |
| Yes | |
| No | |
| Cylindrical | |
| Distilled Water |