missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Right-to-Know Safety-Vented Wash Bottles, GHS Labeling

Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips. Allows dispensing of wash bottle in upright or inverted position.

Supplier:  Bel-Art™ F124320015

 View more versions of this product

Catalog No. 03411739



Get More Bel-Art Lab Products with Purchase
View all promotions
Only null left

Description

Description

Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position

  • Wide mouth 53 mm (2 in.) for easy refilling
  • Translucent LDPE bottles with color-coded caps for easy identification
  • Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word
  • Methyl Ethyl Ketone and Toluene do not include vented valve
  • Assortment is 4 per bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol
  • Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
  • Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word.

Compliance

Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to Workplace Labels

Specifications

Chemical Identifiers

LDPE
Yes
No
Not Autoclavable
Non-sterile
Reusable
Cylindrical

Specifications

32 oz., 1000 mL
32 oz.
LDPE
Non-sterile
Reusable
Wide, Threaded
Safety-vented Wash Bottle
Not Autoclavable
1000 mL
Yellow
2/Pk.
Yes
No
Cylindrical
Sodium Hypochlorite (Bleach)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions