missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ PTFE Policeman

Scrape and spread without scratching

Supplier:  Bel-Art™ H377380000

Numéro de catalogue. 22261767



Il en reste null

Description

Description

  • Inert and chemical resistant
  • With flattened ends
  • Steam autoclavable at 121°C (250°F) for reuse
  • 10 in. (254mm) long
  • 0.3125 in. (7.9mm) diameter

Spécifications

Spécifications

Blunt End
Teflon Lab Policeman
10 in.
0.312 in.
203 x 25 mm
6 mm
254 mm
Autoclavable
For aggressive scraping and spreading
25 mm
8mm
Flattened ends
PTFE
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions