missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Pipette Pump™ Pipetters

Catalog No. 13683D
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
Volume (Metric):
0.2 mL
2 mL
10 mL
25 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric)
13683D Red 25 mL
13683B Blue 2 mL
13683A Yellow 0.2 mL
13683C Green 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13683D Supplier Bel-Art™ Supplier No. 37899 0000
Get Up to $500 in Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides safe, accurate and trouble-free pipetting. Bel Art™ Pipette Pump™ Pipetters utilize an easy-to-use one-handed operation.

  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121ºC (250ºF) for 20 minutes; chuck should not be autoclaved
  • Replacement Silicone Rubber Chuck and Washers are available

Specifications

Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Red
Disposable Reusable
For Use With Pipette
Grip Type Ergonomic
Material Polypropylene
Model Pipette Pump Pipettors
Number of Channels 1
Sterility Non-sterile
Type Pipette
Volume (Metric) 25 mL
For Use With (Equipment) Pipets
Resistance Acid and Alkali
Show More Show Less