missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Mini Sampler Spoons

Catalog No. 03410519
Change view
Click to view available options
Capacity (Metric):
0.05 mL
0.1 mL
0.25 mL
0.5 mL
1 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03410519 0.25 mL
03410518 0.1 mL
03410517 0.05 mL
03410520 0.5 mL
03410521 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03410519 Supplier Bel-Art™ Supplier No. F367210025
Get Up to $500 in Products with Purchase
View all promotions
Edge

Avoid cross contamination by using a different spoon for each sample

  • These disposable mini measures are designed for retrieving small, specific volume samples of powders or similar materials
  • Made from white high-impact polystyrene plastic
  • All sterile products are lot stamped for accurate tracing

Specifications

Sterility Sterile
Autoclavable Not Autoclavable
Capacity (Metric) 0.25 mL
Color White
Disposable Single-use
Product Type Spoon
For Use With (Application) Designed for retrieving small, specific volume samples of powders or similar materials
Material Polystyrene