Learn More
Bel-Art™ Churn Sample Splitter
Provides an efficient method of preparing subsamples of collected water-sediment mixtures from stream flows for water quality analysis
Supplier: Bel-Art™ F378050014
Description
- Polyethylene sample splitter consists of thick walled cylindrical vessel with disc churn
- Center handle extends through cover and locating notch keeps it oriented in one direction
- Small, polypropylene spring-loaded spigot drains subsamples into bottles for delivery to water quality laboratory
- Heavy duty swing type handles supports full or empty churn
- Built-in full open lever stop on spigot ensures reproducible dispensing of subsamples
- Water sediment samples are poured into sample splitter where they are composited into one representative cross section sample of stream flow
- Lid, disc churn and spigot are replaceable
- Intended to meet USGS water sampling protocol
Identifiants chimiques
| Churn Sample Splitter | |
| Polyethylene | |
| Not Autoclavable | |
| Provides an efficient method of preparing subsamples of collected water-sediment mixtures from stream flows for water quality analysis |
Spécifications
| Churn Sample Splitter | |
| Not Autoclavable | |
| 3.6 gal. | |
| Provides an efficient method of preparing subsamples of collected water-sediment mixtures from stream flows for water quality analysis | |
| 30 cm | |
| 30.5 cm |
| Polyethylene | |
| Thick walled cylindrical vessel with disc churn, small polypropylene spring-loaded spigot, built-in full open lever, heavy duty swing type handle | |
| 14 L | |
| 11.8 in. | |
| 12 in. |