missing translation for 'onlineSavingsMsg'
Learn More

Behenic Acid, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012512
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012512 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012512 Supplier Merck Emd Millipore Supplier No. 11909100MG

Behenic acid is a saturated fatty acid with cholesterol-raising ability in humans.

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 112-85-6
Physical Form Neat
Linear Formula CH3(CH2)20 COOH
MDL Number MFCD00002807
Melting Point 72°C to 80°C; 79°C to 81°C
Synonym Docosanoic acid
Quantity 100 mg
Formula Weight 340.58
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less