Electron Microscopy Sciences BEEM Flat Embedding Mold
Numéro de catalogue. 50930536
Molded from polyethylene. The critical surfaces of the mold are transparent, facilitating accurate specimen orientation through bottom illumination. Molds are reusable and each mold is supplied with a protective box.
Numéro de catalogue.
50930536
Fournisseur
Electron Microscopy SciencesCode fournisseur
7090401
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Molded from polyethylene. The critical surfaces of the mold are transparent, facilitating accurate specimen orientation through bottom illumination. Molds are reusable and each mold is supplied with a protective box.