missing translation for 'onlineSavingsMsg'
Learn More

Beclomethasone Dipropionate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012211
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012211 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012211 Supplier Merck Emd Millipore Supplier No. PHR16191G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Beclomethasone Dipropionate is a second generation anti-inflammatory corticosteroid used for the treatment of allergic rhinitis.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 9-8-5534
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C28H37 ClO7
MDL Number MFCD00135613
Synonym Beclomethasone dipropionate; 9-Chloro-11 beta,17,21-trihydroxy-16 beta-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate
Type Certified Reference Material
Quantity 1 g
Formula Weight 521.04
Grade Certified Reference Material