missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Omni International Bead Ruptor 96 25 mL Milling Jars and O-rings Set of 2

Bead Ruptor 96 Milling Jars and O-rings are designed for rapid and efficient disruption of larger samples.
Supplier: Omni International 27004
Description
- 2 stainless steel milling jars and o-rings
- The jars are sealed via a screw mechanism with an o-ring interface.
- Stainless steel jars are ideal for dry grinding or cryomilling applications of very tough material including, bone, seeds, paper and plastics.
Spécifications
| Milling Jars | |
| Stainless Steel | |
| Bead Ruptor 96 | |
| Very tough material including, bone, seeds, paper and plastics |
| Autoclavable | |
| Ideal for dry grinding or cryomilling applications. | |
| 4 to 10 mL |