missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Vitamin B12 Assay Medium

Catalog No. DF0360157
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0360157 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0360157 Supplier BD Supplier No. B236010

For determining vitamin B12 concentration by the microbiological assay technique

Order Info

Prices are for units of one each unless otherwise specified

Certifications

ISO, 9000

Specifications

Certifications/Compliance ISO, 9000
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Vitamin B12 Assay Medium
Product Type Microbiology Media
Formulation Vitamin assay Casamino Acids 12g/L, Dextrose 40g/L, Sodium Acetate 20g/L, L-Cystine 0.2g/L, DL-Tryptophan 0.2g/L, Adenine 20mg/L, Guanine 20mg/L, Uracil 20mg/L, Xanthine 1mg/L, Thiamine Hydrochloride 2mg/L, Riboflavin 2mg/L, Niacin 2mg/L, Calcium Pantothenate 200μg/L, Pyridoxine Hydrochloride 4mg/L, p-Aminobenzoic Acid 200μg/L, Biotin 10μg/L, Folic Acid 100μg/L, Polysorbate 80 2g/L, Dipotassium Phosphate 1g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.4g/L, Sodium Chloride 20mg/L, Ferrous Sulfate 20mg/L, Manganese Sulfate 20mg/L
For Use With (Application) For determining vitamin B12 concentration by the microbiological assay technique
Packaging Type Bottle
Quantity 100 g