missing translation for 'onlineSavingsMsg'
Learn More

BDI Reagent, Ricca Chemical

Catalog No. 9101
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9101 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 9101 Supplier Ricca Chemical Company Supplier No. 9101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Acid Degree Value (Hydrolytic Rancidity Test)

Specifications

Chemical Name or Material BDI Reagent
Packaging Natural Poly Bottle
Name Note for Acid Degree Value (Hydrolytic Rancidity Test)
CAS 10124-56-8, 9002-93-1, 7732-18-5
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Solution
Quantity 4 L
Grade Laboratory
Show More Show Less