missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDE No 77 Solution, 50 μg/mL in isooctane, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| CAS | 93703-48-1 |
| Flash Point | -12°C - Closed Cup |
| Linear Formula | C12H6 Br4 O |
| MDL Number | MFCD00626455 |
| Synonym | 3,3 ′,4,4 ′-Tetrabromodiphenyl ether solution; 3,3 ′,4,4 ′-TetraBDE; PBDE 77 |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 485.79 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 50 μg/mL in isooctane |