missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDE No 154 Solution, 50 μg/mL in isooctane, MilliporeSigma™ Supelco™
Description
Environmental
Spécifications
Spécifications
| CAS | 207122-15-4 |
| Flash Point | -12°C |
| Linear Formula | C12H4 Br6 O |
| MDL Number | MFCD01657488 |
| Synonym | 2,2 ′,4,4 ′,5,6 ′-Hexabromodiphenyl ether solution; PBDE 154 |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 643.58 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 50 μg/mL in isooctane |