missing translation for 'onlineSavingsMsg'
Learn More

BD Veritor™ System Flu A+B Kit

Catalog No. B256041
Change view
Click to view available options
Quantity:
30 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B256041 30 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B256041 Supplier BD Supplier No. 256041

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Rapid Detection of Flu A+B

  • Easy sample processing - Prefilled unitized tube facilitates workflow
  • Ready in minutes - Test device is ready to insert into reader 10 minutes after sample is added
  • Insert and read - Simple one-touch button readies the reader for test device insertion
  • Objective results - Once the test device is inserted in the reader, an objective, digital test result is displayed in 10 seconds

Specifications

Description Flu A + B Test Kit (For laboratory use)
Clia Complexity Moderate
Detectable Analytes Influenza A and B
Sample Type Nasopharyngeal swabs, nasopharyngeal aspirate and nasopharyngeal washes
Type Flu A + B Test Kit
For Use With (Application) Detection of Flu A + B
Includes Lateral flow test strips, pipettes, Flu A+/B- control swab, Flu B+/A- control swab, RV reagent C in tubes
Packaging Type 1 Kit
Quantity 30 Assays
Test Time 10 min.

For in vitro Diagnostic Use.