Learn More
BD Vacutainer™ Luer Adapters
Catalog No. 0266527
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
0266527 Sterile
1 options
Catalog No. 0266527 Supplier BD Supplier No. 367290
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Especially useful when intravenous fluids or injections are to follow blood withdrawal
  • Designed to allow the attachment of a variety of Luer slip devices (disposable or reusable hypodermic needles, IV catheters and blood collection sets) to BD Vacutainer holder using a single venipuncture
  • Sterile and disposable
  • After blood collection, needle or catheter is left in place for additional procedures, reducing further patient trauma
Specifications
Disposable Single-use
Sterility Sterile
For Use With (Application) Useful when intravenous fluids or injections are to follow Blood withdrawal
Product Type Luer Adapter
For Use With (Equipment) Female luer connection or needleless IV site designed for luer lock access, and the Bard EZ-Lok foley catheter sampling port