Learn More
BD Syringe with Slip (Luer) Tips (Without Needle)
Description
- Single-use with plastic barrels, plungers and tips
- Nonsterile syringes may be irradiated, autoclaved, or gas sterilized out of polybag
Specifications
Specifications
| Product Type | Syringe |
| Graduations | 1/5 mL |
| Needle Point Number | No Needle Included |
| Volume (Metric) | 5 mL |
| Connection Type | Leur Slip |
| Graduated | Yes |
| Sterility | Non-sterile |