Learn More
BD RODAC™ SNAP LID Sabouraud Dextrose Agar (SDA) with Lecithin and Polysorbate 80 (LP80)
Numéro de catalogue. p-4364755
Change view
Click to view available options
Quantity:
10/Pk.
Packaging Type:
VHP Resistant Triple Wrapping Isolator Pack
Numéro de catalogue. Quantity Packaging Type
B215270 10/Pk. VHP Resistant Triple Wrapping Isolator Pack
1 options
Numéro de catalogue. B215270 Fournisseur BD Code fournisseur 215270
Il en reste null
The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.
The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:
  • Withstands an accidental drop from the height of a bench-top workspace
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage
Spécifications
Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type Sabouraud Dextrose Agar with Lecithin and Polysorbate 80
Sterility Sterile
Includes Rodac™ snap lid
Packaging Type VHP Resistant Triple Wrapping Isolator Pack
Quantity 10/Pk.