Learn More
BD BBL™ Gram Slide
Catalog No. B231401
Change view
Click to view available options
Quantity:
50 Pk.
Catalog No. Quantity
B231401 50 Pk.
1 options
Catalog No. B231401 Supplier BD Supplier No. 231401
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used for evaluating and controlling the quality of Gram stain reagents and techniques. BD Difco™ BBL™ Gram Slide enables easy evaluations.
  • Slide format
  • Used for evaluating: Gram stain reagents 
  • 50 slides
Specifications
Content And Storage 30°C
Quantity 50 Pk.