missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Prepared Bottled Media: Tryptic Soy Broth
Description
Ready for immediate use
Spécifications
Spécifications
| Content And Storage | 5°C to 25°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Tryptic Soy Broth |
| Packaging Type | Bottle |
| Quantity | 20 mL |