Learn More
BD Microtainer™ MAP Microtube for Automated Process with K2EDTA and BD Microgard™ Closure
Catalog No. 22253270
Change view
Click to view available options
Capacity (Metric):
250 to 500 μL
Additive:
1.0 mg K2 EDTA
Catalog No. Capacity (Metric) Additive
22253270 250 to 500 μL 1.0 mg K2 EDTA
1 options
Catalog No. 22253270 Supplier BD Supplier No. 363706
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed for the collection, transport and automated processing of skin puncture samples from infants, children, geriatric, oncology and critical care patients
The BD Microtainer MAP tube is designed to improve turnaround time, safety and accuracy of capillary blood collection and testing in patients with limited blood volume such as infants, children, oncology patients and the elderly.
  • Streamlines operations by allowing for automated processing of your micro-collection hematology samples
  • Enhances patient safety by accommodating full-size patient identification labels at the time of collection
  • Improves workflow with one-piece design, no parts to assemble, no contaminated parts to discard
  • Latex-free, disposable and nonsterile
Specifications
Color Clear
Product Type MAP MicroTube for Automated Process
Additive 1.0 mg K2 EDTA
Sterility Non-sterile
Capacity (Metric) 250 to 500 μL
Closure Included Yes
Closure Color Lavender