missing translation for 'onlineSavingsMsg'
Learn More

BD GasPak™ EZ Gas Generating Systems and Supplies: Pouch Systems

Catalog No. B260683
Change view
Click to view available options
Type:
EZ Anaerobe Pouch System
EZ CampyPouch System
EZ CO2 Pouch System
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
B260683 EZ Anaerobe Pouch System
B260684 EZ CO2 Pouch System
B260685 EZ CampyPouch System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B260683 Supplier BD Supplier No. 260683
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in producing anaerobic, microaerophilic, or CO2-enriched environments. BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies provide catalyst-free convenience.

  • One-step resealable pouches
  • The pouches are specially designed to maximize the preservation of the desired environment throughout incubation
  • This heat-free technology minimizes condensate, allowing excellent bacterial colony visualization though the pouch

Specifications

Type EZ Anaerobe Pouch System
Closure Type Ziplock
Format Resealable Pouches
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
For Use With (Equipment) BD GasPak EZ Small Incubation Container
Includes Resealable pouches with indicator
Product Line GasPak
Quantity 20 Sachets