missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Riboflavin Assay Medium

Catalog No. DF0325151
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0325151 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0325151 Supplier BD Supplier No. 232510

For determining riboflavin concentration by the microbiological assay technique

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

AOAC

Specifications

Certifications/Compliance AOAC
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Riboflavin Assay Medium
Product Type Riboflavin Assay Medium
Formulation Dextrose 20g/L, Sodium Acetate 15g/L, Vitamin assay Casamino Acids 10g/L, Dipotassium Phosphate 1g/L, Monopotassium Phosphate 1g/L, L-Asparagine 0.6g/L, DL-Tryptophan 0.2g/L, L-Cystine 0.2g/L, Magnesium Sulfate USP 0.4g/L, Adenine Sulfate 20mg/L, Guanine
For Use With (Application) Microbiological assay of Riboflavin
Packaging Type Bottle
Quantity 100 g