missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Mueller Hinton Agar
Description
- Dehydrated
- Culture Media
- Mueller Hinton Agar
Specifications
Specifications
| Certifications/Compliance | BAM, BS12, CCAM, CLSI, CMPH2, ISO, MCM9 |
| Content And Storage | 2°C to 30°C |
| Form | Dehydrated |
| Media Type | Mueller Hinton Agar |
| Product Type | Mueller Hinton Medium |
| Formulation | Beef extract 2g/L, Acid digest of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L |
| For Use With (Application) | For antimicrobial disc diffusion susceptibility testing of common, rapidly growing bacteria by the Bauer-Kirby method,1-3 as standardized by the clinical and laboratory standards institute |
| Packaging Type | Bottle |
| Quantity | 500 g |