missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: M 17 Broth

Catalog No. DF1856174
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF1856174 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF1856174 Supplier BD Supplier No. 218561
Edge

Selective isolation and enumeration of streptococci. BD Difco™ Dehydrated Culture Media: M 17 Broth is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • M 17 Broth

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Certifications

MPF, ISO

Specifications

Certifications/Compliance COMPF, ISO
Content And Storage Refrigerate
Form Dehydrated
Media Type M17 Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 5g/L, Soy Peptone 5g/L, Beef extract 5g/L, Yeast extract 2.5g/L, Ascorbic Acid 0.5g/L, Magnesium Sulfate 0.25g/L, Disodium-beta-glycerophosphate 19g/L
For Use With (Application) Isolating lactic streptococci from yogurt, cheese starters and other dairy product
Packaging Type Bottle
Quantity 500 g