missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Brewer Anaerobic Agar

Catalog No. DF0279175
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0279175 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0279175 Supplier BD Supplier No. 227920
Only null left

For cultivation of anaerobic and microaerophilic organisms

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type Brewer Anaerobic Agar
Product Type Agar
Formulation Pancreatic digest of Casein 5g/L, Proteose Peptone No. 3 10g/L, Yeast extract 5g/L, Dextrose 10g/L, Sodium Chloride 5g/L, Agar 20g/L, Sodium Thioglycollate 2g/L, Sodium Formaldehyde Sulfoxylate 1g/L, Resazurin 2mg/L
For Use With (Application) Cultivation of anaerobic and microaerophilic organisms
Packaging Type Bottle
Quantity 500 g