missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Bismuth Sulfite Agar

Catalog No. DF0073173
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0073173 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0073173 Supplier BD Supplier No. 273300
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For selective isolation of Salmonella, particularly Salmonella typhi, from food and clinical specimens

Specifications

Certifications/Compliance ISO, 9000
Content And Storage <30°C
Form Dehydrated
Media Type Bismuth Sulfite Agar
Product Type Agar
Formulation Beef extract 5g/L, Peptone 10g/L, Dextrose 5g/L, Disodium Phosphate 4g/L, Ferrous Sulfate 0.3g/L, Bismuth Sulfite Indicator 8g/L, Agar 20g/L, Brilliant green 25mg/L
For Use With (Application) Isolation of Enterococcus faecalis 29212, Escherichia coli 25922, Salmonella enterica subspecies enterica serotype Typhi 19430, Salmonella enterica subspecies enterica serotype Typhimurium 14028
Packaging Type Bottle
Quantity 500 g