Learn More
BD Cannulas
Catalog No. 361042703
Change view
Click to view available options
Product Type:
Vial Access Cannula
Catalog No. Product Type
361042703 Vial Access Cannula
1 options
Catalog No. 361042703 Supplier BD Supplier No. 303367
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Needle Gauge 15 G
Product Type Vial Access Cannula
For Use With (Equipment) Interlink Injection Sites
Sterility Sterile