Learn More
BD BBL™ SEPTI-CHEK™ Blood Culture System: Sub/Venting units
Catalog No. B4371056
Change view
Click to view available options
Quantity:
100/Pk.
Catalog No. Quantity
B4371056 100/Pk.
1 options
Catalog No. B4371056 Supplier BD Supplier No. 271056
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed for rapid qualitative detection, isolation, and differentiation of pathogens from blood
  • Closed, biphasic system
  • Risk of contamination and sample mix-up is minimized
Specifications
Product Type Sub/Venting Unit
For Use With (Equipment) Septi check™ culture bottle
Quantity 100/Pk.