Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Gentamicin
Catalog No. B31299
Change view
Click to view available options
Quantity:
1 Cartridge (50 discs)
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31299 10/Pk.
B31227 1 Cartridge (50 discs)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31299 Supplier BD Supplier No. 231299
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Antimicrobial Agents Gentamicin
Concentration 10 μg
Packaging Type Disc
Quantity 10/Pk.