Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Ampicillin4, AM-10
Catalog No. B30705
Change view
Click to view available options
Quantity:
1/Pk.
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30705 1/Pk.
B31264 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30705 Supplier BD Supplier No. 230705
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Employs Bauer-Kirby disc diffusion testing method
  • Use in self-tamping dispensers
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
  • Cartridges of 50 discs each are sold individually or in packs of 10
Specifications
Antimicrobial Agents Ampicillin
Concentration 10 μg
Packaging Type Disc
Quantity 1/Pk.