Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefixime1414
Catalog No. B31664
Change view
Click to view available options
Concentration:
5 μg
Catalog No. Concentration
B31664 5 μg
1 options
Catalog No. B31664 Supplier BD Supplier No. 231664
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Antimicrobial Agents Cefixime14
Concentration 5 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro diagnostic use