Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefepime13
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Supplier:  BD 231696
Numéro de catalogue. B4331696
Il en reste null
Description
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Spécifications
Spécifications
Cefepime13
30 μg
Disc
10 Pack
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
Sécurité et manipulation
Sécurité et manipulation
missing translation for 'shelfLife' : 548 Days
FDS
Documents
Documents
Promotions
Promotions
For in vitro diagnostic use