missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Replacement Reagents for Gram Stain Kits

Numéro de catalogue. B4312545
Change view
Click to view available options
Quantity:
3.8 L
4 x 250 mL
Type:
Basic Fuchsin
Gram Iodine
Gram Iodine (Stabilized)
Safranin
Stain Type:
Basic Fuchsin
Iodine
Safranin
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Type Stain Type
B4312545 3.8 L Basic Fuchsin Basic Fuchsin
B4312531 4 x 250 mL Safranin Safranin
B4312532 3.8 L Safranin Safranin
B4312542 4 x 250 mL Gram Iodine Iodine
B4312543 3.8 L Gram Iodine (Stabilized) Iodine
B4312544 4 x 250 mL Basic Fuchsin Basic Fuchsin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. B4312545 Fournisseur BD Code fournisseur 212545
Il en reste null

For BBL Gram Stain Kits

  • Complete stain kits reduce the mess associated with Gram staining procedures
  • Break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills
  • Color-coded labels allow quick identification of reagents
  • Stabilized Iodine Solution requires no mixing and has a long shelf life

Order Info

Available in packs of four 250mL bottles or individual one-gallon bottles; sales of B12542 are restricted from California

Spécifications

Type Basic Fuchsin
Quantity 3.8 L
Certifications/Compliance CE
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Storage Requirements 15° to 30°C
Stain Type Basic Fuchsin